missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Antibody (5G12C4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28893-0.025ml
This item is not returnable.
View return policy
Description
ITK Monoclonal specifically detects ITK in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ITK | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| ITK | |
| Purified recombinant fragment of ITK(aa2-110) expressed in E. Coli. | |
| 0.025 mL | |
| Protein Kinase | |
| 3702 | |
| Human | |
| Ascites |
| Western Blot | |
| 5G12C4 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| EC 2.7.10, EC 2.7.10.2, EMTMGC126257, homolog of mouse T-cell itk/tsk, IL2-inducible T-cell kinase, Kinase EMT, LYKMGC126258, PSCTK2, T-cell-specific kinase, tyrosine-protein kinase ITK/TSK, Tyrosine-protein kinase Lyk | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific for ITK. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction