missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Antibody (5G12C4), Novus Biologicals™
Mouse Monoclonal Antibody
168.00€
Specifications
| Antigen | ITK |
|---|---|
| Clone | 5G12C4 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
ITK Monoclonal specifically detects ITK in Human samples. It is validated for Western Blot, ELISA.Specifications
| ITK | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| EC 2.7.10, EC 2.7.10.2, EMTMGC126257, homolog of mouse T-cell itk/tsk, IL2-inducible T-cell kinase, Kinase EMT, LYKMGC126258, PSCTK2, T-cell-specific kinase, tyrosine-protein kinase ITK/TSK, Tyrosine-protein kinase Lyk | |
| ITK | |
| IgG1 | |
| Unpurified | |
| Specific for ITK. |
| 5G12C4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| Ascites with 0.03% Sodium Azide | |
| 3702 | |
| Purified recombinant fragment of ITK(aa2-110) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title