missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 3/CD49c Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
325.00€
Specifications
| Antigen | Integrin alpha 3/CD49c |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Chicken |
Description
Integrin alpha 3/CD49c Polyclonal specifically detects Integrin alpha 3/CD49c in Human, Rat samples. It is validated for Western Blot.Specifications
| Integrin alpha 3/CD49c | |
| Polyclonal | |
| Chicken | |
| Human | |
| P26006 | |
| 3675 | |
| The immunogen for this antibody was a sequence from 1015a.a. to 1051a.a. from Integrin alpha-3(a 1051a.a. long protein) | |
| Affinity Purified | |
| Integrin alpha 3 |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with No Preservative | |
| antigen identified by monoclonal J143, CD49 antigen-like family member C, CD49c, CD49c antigen, FLJ34631, FLJ34704, FRP-2, Galactoprotein B3, GAP-B3, GAPB3CD49C, integrin alpha-3, integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor), MSK18, VCA-2, very late activation protein 3 receptor, alpha-3 subunit, VL3A, VLA-3 subunit alpha, VLA3a | |
| ITGA3 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title