missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 3/CD49c Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75266-0.025mg
This item is not returnable.
View return policy
Description
Integrin alpha 3/CD49c Polyclonal specifically detects Integrin alpha 3/CD49c in Human, Rat samples. It is validated for Western Blot.
Specifications
| Integrin alpha 3/CD49c | |
| Polyclonal | |
| PBS with No Preservative | |
| antigen identified by monoclonal J143, CD49 antigen-like family member C, CD49c, CD49c antigen, FLJ34631, FLJ34704, FRP-2, Galactoprotein B3, GAP-B3, GAPB3CD49C, integrin alpha-3, integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor), MSK18, VCA-2, very late activation protein 3 receptor, alpha-3 subunit, VL3A, VLA-3 subunit alpha, VLA3a | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 3675 | |
| Human |
| Western Blot | |
| Unconjugated | |
| P26006 | |
| ITGA3 | |
| The immunogen for this antibody was a sequence from 1015a.a. to 1051a.a. from Integrin alpha-3(a 1051a.a. long protein) | |
| 0.025 mg | |
| Primary | |
| Integrin alpha 3 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction