Learn More
Instrument Check Standards for ICP/MS, Set of 5, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
5 x 125 mL
Unit Size:
1 set
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Instrument Check Standards for ICP-MS
- Set of 5
- 125 mL Volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Instrument Check Standards, Set of 5 |
| Name Note | Set of 5 |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | ICP-MS |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Solution Type | Standard Solution |
| Type | ICP/ICP-MS Standard |
| Quantity | 5 x 125 mL |
| Grade | ICP/ICP-MS |
| Show More |
Product intended for laboratory use only