missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H1N1 M1 Antibody - (A/Puerto Rico/8/34/Mount Sinai), Novus Biologicals™

Rabbit Polyclonal Antibody

299.00€ - 507.00€

Specifications

Antigen Influenza A H1N1 M1
Dilution Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18650277
View Documents
Novus Biologicals
NBP3-12721-50ul
50 μg
299.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18631867
View Documents
Novus Biologicals
NBP3-12721-100ul
100 μg
507.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Influenza A H1N1 M1 Polyclonal antibody specifically detects Influenza A H1N1 M1 in Influenza A virus H1N1 samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Influenza A H1N1 M1
Western Blot, ELISA
Unconjugated
Rabbit
Influenza A virus H1N1
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 M1 (Accession#: NP_040978.1; Ser2-Lys252).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
0.2 um filtered solution in PBS
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.