missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 M1 Antibody - (A/Puerto Rico/8/34/Mount Sinai), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12721-100ul
This item is not returnable.
View return policy
Description
Influenza A H1N1 M1 Polyclonal antibody specifically detects Influenza A H1N1 M1 in Influenza A virus H1N1 samples. It is validated for Western Blot, ELISA
Specifications
| Influenza A H1N1 M1 | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Influenza A virus H1N1 | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Influenza A H1N1 M1 (Accession#: NP_040978.1; Ser2-Lys252). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction