missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
205.00€ - 472.00€
Specifications
| Antigen | IL-6 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 2 μg/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696151
|
Novus Biologicals
NBP2-78132 |
0.1 mg |
472.00€
0.01mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674829
|
Novus Biologicals
NBP2-78132SS |
0.025 mg |
205.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-6 Polyclonal antibody specifically detects IL-6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| IL-6 | |
| Western Blot 1 - 2 μg/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
| Partial recombinant mouse IL-6 protein [UniProt P08505] | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
| PBS | |
| 3569 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title