missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-34 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€
Specifications
| Antigen | IL-34 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.25 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
IL-34 Polyclonal specifically detects IL-34 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| IL-34 | |
| Western Blot 0.25 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| ACD13474 | |
| 146433 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human IL-34. The immunogen is located within amino acids 30 - 80 of IL-34. Amino Acid Squence: LRDKLQYRSRLQYMKH | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| C16orf77, IL-34, interleukin 34 | |
| IL34 | |
| IgG | |
| Affinity Purified | |
| 42-48 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title