missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-34 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76357-0.025mg
This item is not returnable.
View return policy
Description
IL-34 Polyclonal specifically detects IL-34 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| IL-34 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C16orf77, IL-34, interleukin 34 | |
| Rabbit | |
| 42-48 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.25 μg/mL, ELISA 1:100-1:2000 | |
| ACD13474 | |
| IL34 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human IL-34. The immunogen is located within amino acids 30 - 80 of IL-34. Amino Acid Squence: LRDKLQYRSRLQYMKH | |
| Affinity Purified | |
| RUO | |
| 146433 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction