missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB600-1443-0.025mg
This item is not returnable.
View return policy
Description
IL-12 Polyclonal specifically detects IL-12 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IL-12 | |
| Polyclonal | |
| Western Blot 1-2 μg/mL, ELISA 0.5-1 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Neutralization | |
| P43431 | |
| IL12A | |
| Purified, insect cell line Sf21-derived, recombinant mouse IL-12 | |
| 0.025 mg | |
| Immunology, Innate Immunity | |
| 3592 | |
| To one vial of lyophilized powder, add 1 mL of 0.2 mm filtered phosphate buffered saline to produce a 1 mg/mL stock solution of the antibody. If aseptic technique is used, no further filtration should be needed for use in cell cμLture environments. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Lyophilized from 0.2 um-filtered solution in phosphate buffered saline containing carbohydrates. with No Preservative | |
| CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| Goat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Based on ELISA, the antibody shows ∼10% cross-reactivity with recombinant human IL-12 and <5% cross-reactivity with recombinant porcine IL-12. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction