missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
215.00€
Specifications
| Antigen | IL-12 |
|---|---|
| Dilution | Western Blot 1-2 μg/mL, ELISA 0.5-1 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Neutralization |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IL-12 Polyclonal specifically detects IL-12 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| IL-12 | |
| Western Blot | |
| Unconjugated | |
| Goat | |
| Immunology, Innate Immunity | |
| Lyophilized from 0.2 um-filtered solution in phosphate buffered saline containing carbohydrates. with No Preservative | |
| CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| IL12A | |
| IgG | |
| Protein G purified | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot 1-2 μg/mL, ELISA 0.5-1 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Neutralization | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| P43431 | |
| 3592 | |
| Purified, insect cell line Sf21-derived, recombinant mouse IL-12 | |
| Primary | |
| To one vial of lyophilized powder, add 1 mL of 0.2 mm filtered phosphate buffered saline to produce a 1 mg/mL stock solution of the antibody. If aseptic technique is used, no further filtration should be needed for use in cell cμLture environments. | |
| Based on ELISA, the antibody shows ∼10% cross-reactivity with recombinant human IL-12 and <5% cross-reactivity with recombinant porcine IL-12. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title