missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Antibody (JES3-9D7) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
242.00€ - 543.00€
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | JES3-9D7 |
| Concentration | 0.5 mg/mL |
| Applications | Western Blot, ELISA, ELISA Capture (Matched Antibody Pair) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625771
|
Novus Biologicals
NBP2-77379-0.5mg |
0.5 mg |
543.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633311
|
Novus Biologicals
NBP2-77379-0.1mg |
0.1 mg |
242.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human, Cynomolgus Monkey, Macaque, Virus samples. It is validated for Western Blot, ELISA, ELISA Capture (Matched Antibody Pair)Specifications
| IL-10 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Cynomolgus Monkey, Macaque, Virus | |
| CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| COS-expressed human IL-10 | |
| Primary | |
| Store at 4°C. Do not freeze. |
| JES3-9D7 | |
| Western Blot, ELISA, ELISA Capture (Matched Antibody Pair) | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 1.0 mL of borate buffered saline, pH 8.2. | |
| 3586 | |
| IgG1 κ | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title