Learn More
IL-10 Antibody (JES3-9D7) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-77379-0.5mg
 View more versions of this product
Product Code. 18625771
  • 543.00€ / 0.50mg
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human, Cynomolgus Monkey, Macaque, Virus samples. It is validated for Western Blot, ELISA, ELISA Capture (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-10
Monoclonal
0.5 mg/mL
1.0 mL of borate buffered saline, pH 8.2.
Rat
Protein G purified
RUO
Primary
Human, Cynomolgus Monkey, Macaque, Virus
Purified
Western Blot, ELISA, ELISA Capture (Matched Antibody Pair)
JES3-9D7
Unconjugated
CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
COS-expressed human IL-10
0.5 mg
Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
3586
Store at 4°C. Do not freeze.
IgG1 κ