missing translation for 'onlineSavingsMsg'
Learn More

IgG (H, L) Chicken anti-Cotton Rat, Polyclonal , Abnova™

Product Code. 16048197
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16048197 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16048197 Supplier Abnova Supplier No. PAB29752.1mg
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken polyclonal antibody raised against cotton rat IgG (H&L).

Specifications

Antigen IgG (H, L)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against cotton rat IgG (H&L).
Dilution ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Chicken
Immunogen Cotton rat IgG (H&L) whole molecule.
Purification Method Antigen Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The affinity purified anti Cotton rat IgG reacts with immunoglobulins and the papain derived Fc and Fab fragments by IEP and gel diffusion techniques. It also reacts with rat, gerbil and hamster IgG.
Target Species Cotton Rat
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.