missing translation for 'onlineSavingsMsg'
Learn More

IgG (H, L) Chicken anti-Cotton Rat, HRP, Polyclonal , Abnova™

Product Code. 16058197
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16058197 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16058197 Supplier Abnova Supplier No. PAB29753.1mg
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken polyclonal antibody raised against cotton rat IgG (H&L).

Specifications

Antigen IgG (H, L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Description Chicken polyclonal antibody raised against cotton rat IgG (H&L).
Dilution ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.5-5 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM HEPES-Saline with stabilizer (0.002% Thimerosal)
Host Species Chicken
Immunogen Cotton rat IgG (H&L) whole molecule.
Purification Method Antigen Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The antibody reacts with all classes of Cotton Rat immunoglobulins. By IEP, gel diffusion and ELISA. It also reacts with gerbil, rat and hamster IgG.
Target Species Cotton Rat
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.