missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Antibody (11-26c.2a), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
589.00€
Specifications
| Antigen | IgD |
|---|---|
| Clone | 11-26c.2a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
Description
IgD Monoclonal specifically detects IgD in Mouse samples. It is validated for Flow Cytometry.Specifications
| IgD | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Rat | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 3495 | |
| This antibody was generated by immunizing host with mouse IgD | |
| Primary | |
| Store at 4°C in the dark. |
| 11-26c.2a | |
| Monoclonal | |
| Purified | |
| Mouse | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| IGHD | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title