missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Antibody (11-26c.2a), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-49000AF647
This item is not returnable.
View return policy
Description
IgD Monoclonal specifically detects IgD in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IgD | |
| Monoclonal | |
| Alexa Fluor 647 | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 11-26c.2a | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IGHD | |
| This antibody was generated by immunizing host with mouse IgD | |
| 0.5 mL | |
| 3495 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction