missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
279.00€ - 760.00€
Specifications
| Antigen | IFI27 |
|---|---|
| Dilution | ELISA 1:10000-1:1000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18255984
|
Novus Biologicals
NB300-709-0.025MG |
0.025 mg |
279.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18204085
|
Novus Biologicals
NB300-709-0.05MG |
0.05 mg |
463.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18281480
|
Novus Biologicals
NB300-709 |
0.1 mg |
760.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFI27 Polyclonal specifically detects IFI27 in Human, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| IFI27 | |
| ELISA | |
| Unconjugated | |
| Human | |
| 3429 | |
| Synthetic peptide. | |
| Primary |
| ELISA 1:10000-1:1000 | |
| Polyclonal | |
| Rabbit | |
| FAM14D, Interferon alpha-induced 11.5 kDa protein, interferon alpha-inducible protein 27, mitochondrial, interferon, alpha-inducible protein 27,2310061N23Rik, Interferon-stimulated gene 12a protein, ISG12, ISG12(a), ISG12A, P27 | |
| IFI27 | |
| IgG | |
| IFI27 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title