missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-709
This item is not returnable.
View return policy
Description
IFI27 Polyclonal specifically detects IFI27 in Human, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| IFI27 | |
| Polyclonal | |
| ELISA 1:10000-1:1000 | |
| FAM14D, Interferon alpha-induced 11.5 kDa protein, interferon alpha-inducible protein 27, mitochondrial, interferon, alpha-inducible protein 27,2310061N23Rik, Interferon-stimulated gene 12a protein, ISG12, ISG12(a), ISG12A, P27 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| IFI27 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| IFI27 | |
| Synthetic peptide. | |
| 0.1 mg | |
| Angiogenesis, Cancer | |
| 3429 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction