missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TNF RII/TNFRSF1B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
242.00€ - 2155.00€
Spezifikation
| Antigen | TNF RII/TNFRSF1B |
|---|---|
| Clone | 22210 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.5-1.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
18729708
![]() |
R&D Systems
MAB726-SP |
25 μg |
242.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18717819
![]() |
R&D Systems
MAB726-100 |
100 μg |
541.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773687
![]() |
R&D Systems
MAB726-500 |
500 μg |
2155.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Spezifikation
| TNF RII/TNFRSF1B | |
| Western Blot 1 ug/mL, Neutralization 0.5-1.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P20333 | |
| 7133 | |
| E. coli-derived recombinant human TNF RII/TNFRSF1B Leu23-Asp257 Accession # P20333 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TNF RII in ELISAs and Western blots. In ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) TNF RI, recombinant mouse (rm) TNF RI, rmTNF RII, rmTNF-alpha , rhTNF-alpha , or rhTNF-beta . In Western blots, this antibody does not cross-react with rhTNF RI, rmTNF RI, or rmTNF RII. |
| 22210 | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| TNFRSF1B | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts