missing translation for 'onlineSavingsMsg'
Learn More

Human TNF RII/TNFRSF1B Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18773687
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18773687 500 μg 500µg
18729708 25 μg 25µg
18717819 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18773687 Supplier R&D Systems Supplier No. MAB726500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 9 publications

TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF RII/TNFRSF1B
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 22210
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.5-1.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P20333
Gene Alias CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Gene Symbols TNFRSF1B
Host Species Mouse
Immunogen E. coli-derived recombinant human TNF RII/TNFRSF1B Leu23-Asp257 Accession # P20333
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7133
Test Specificity Detects human TNF RII in ELISAs and Western blots. In ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) TNF RI, recombinant mouse (rm) TNF RI, rmTNF RII, rmTNF-alpha , rhTNF-alpha , or rhTNF-beta . In Western blots, this antibody does not cross-react with rhTNF RI, rmTNF RI, or rmTNF RII.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.