Learn More
Abnova™ Human SLC12A1 Full-length ORF (ENSP00000331550, 1 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006557-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henle's loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene. [provided by RefSeq]
Sequence: MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSISGPKVNRPSLLEIHEQLAKNVAVTPSSADRVANGDGIPGDEQAENKEDDQAGVVKFGWVKGVLVRCMLNIWGVMLFIRLSWIVGEAGIGLGVLIILLSTMVTSITGLSTSAIATNGFVRGGGAYYLISRSLGPEFGGSIGLIFAFANAVAVAMYVVGFAETVVDLLKESDSMMVDPTNDIRIIGSITVVILLGISVAGMEWEAKAQVILLVILLIAIANFFIGTVIPSNNEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANISGDLEALRKQGASPLPQGAQSRRRRQTSLISpecifications
ENSP00000331550 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BSC1/MGC48843/NKCC2 | |
SLC12A1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6557 | |
SLC12A1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSISGPKVNRPSLLEIHEQLAKNVAVTPSSADRVANGDGIPGDEQAENKEDDQAGVVKFGWVKGVLVRCMLNIWGVMLFIRLSWIVGEAGIGLGVLIILLSTMVTSITGLSTSAIATNGFVRGGGAYYLISRSLGPEFGGSIGLIFAFANAVAVAMYVVGFAETVVDLLKESDSMMVDPTNDIRIIGSITVVILLGISVAGMEWEAKAQVILLVILLIAIANFFIGTVIPSNNEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANISGDLEALRKQGASPLPQGAQSRRRRQTSLI | |
RUO | |
SLC12A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |