missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC12A1 Full-length ORF (ENSP00000331550, 1 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | ENSP00000331550 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6557 |
Molecular Weight (g/mol) | 72.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16190802
|
Abnova™
H00006557-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16100812
|
Abnova™
H00006557-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henle's loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene. [provided by RefSeq]
Sequence: MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSISGPKVNRPSLLEIHEQLAKNVAVTPSSADRVANGDGIPGDEQAENKEDDQAGVVKFGWVKGVLVRCMLNIWGVMLFIRLSWIVGEAGIGLGVLIILLSTMVTSITGLSTSAIATNGFVRGGGAYYLISRSLGPEFGGSIGLIFAFANAVAVAMYVVGFAETVVDLLKESDSMMVDPTNDIRIIGSITVVILLGISVAGMEWEAKAQVILLVILLIAIANFFIGTVIPSNNEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANISGDLEALRKQGASPLPQGAQSRRRRQTSLISpecifications
ENSP00000331550 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSLNNSSNVFLDSVPSNTNRFQVSVINENHESSAAADDNTDPPHYEETSFGDEAQKRLRISFRPGNQECYDNFLQSGETAKTDASFHAYDSHTNTYYLQTFGHNTMDAVPKIEYYRNTGSISGPKVNRPSLLEIHEQLAKNVAVTPSSADRVANGDGIPGDEQAENKEDDQAGVVKFGWVKGVLVRCMLNIWGVMLFIRLSWIVGEAGIGLGVLIILLSTMVTSITGLSTSAIATNGFVRGGGAYYLISRSLGPEFGGSIGLIFAFANAVAVAMYVVGFAETVVDLLKESDSMMVDPTNDIRIIGSITVVILLGISVAGMEWEAKAQVILLVILLIAIANFFIGTVIPSNNEKKSRGFFNYQASIFAENFGPRFTKGEGFFSVFAIFFPAATGILAGANISGDLEALRKQGASPLPQGAQSRRRRQTSLI | |
RUO | |
SLC12A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6557 | |
SLC12A1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BSC1/MGC48843/NKCC2 | |
SLC12A1 | |
Recombinant | |
wheat germ expression system |