missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Wnt-3a Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
250.00€ - 472.00€
Specifications
| Antigen | Wnt-3a |
|---|---|
| Clone | 217804.2R |
| Dilution | Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
18709578
![]() |
R&D Systems
MAB13242-SP |
25 μg |
250.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18746669
![]() |
R&D Systems
MAB13242-100 |
100 μg |
472.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
Wnt-3a Monoclonal specifically detects Wnt-3a in Human, Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Especificaciones
| Wnt-3a | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A, Wnt3a | |
| E.coli-derived recombinant mouse Wnt-3a Ser36-Gln75, Trp219-Arg269 Accession # P27467 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Wnt-3a in direct ELISAs. |
| 217804.2R | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| P27487 | |
| 89780 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto