missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Wnt-3a Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB13242-100
This item is not returnable.
View return policy
Description
Wnt-3a Monoclonal specifically detects Wnt-3a in Human, Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Wnt-3a | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A, Wnt3a | |
| E.coli-derived recombinant mouse Wnt-3a Ser36-Gln75, Trp219-Arg269 Accession # P27467 | |
| 100 μg | |
| Primary | |
| Detects mouse Wnt-3a in direct ELISAs. | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 217804.2R | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/mL, CyTOF-ready | |
| P27487 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 89780 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction