missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat alpha Tubulin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
210.00€ - 507.00€
Specifications
| Antigen | alpha Tubulin |
|---|---|
| Clone | 961216 |
| Dilution | Western Blot 0.05 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18781199
![]() |
R&D Systems
MAB9344-SP |
25 μg |
210.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18750028
![]() |
R&D Systems
MAB9344-100 |
100 μg |
507.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| alpha Tubulin | |
| Western Blot 0.05 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat | |
| Q71U36 | |
| 7846 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 961216 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA1A, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-Alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| Human TUBA1A synthetic peptide Accession # Q71U36 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human alpha Tubulin in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title