missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse FoxP3 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

245.00€ - 555.00€

Specifications

Antigen FoxP3
Clone 1054C
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18784557
View Documents
R&D Systems
FPK8214A-025
25 Tests
245.00€
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18730837
GREENER_CHOICE
View Documents
R&D Systems
FPK8214A-100
100 Tests
555.00€
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

FoxP3
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Rabbit
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
FOXP3
IgG
Affinity Purified
Detects human and mouse FoxP3 in flow cytometry.
1054C
Flow Cytometry
APC
Human, Mouse
50943
E. coli-derived recombinant human FoxP3 Met1-Leu71
Primary
Store the unopened product at 2 to 8 degreesC. Do not use past expiration date. Protect from light.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.