missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse FoxP3 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems FPK8214A-025
213.35 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| FoxP3 | |
| Monoclonal | |
| APC | |
| AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Detects human and mouse FoxP3 in flow cytometry. | |
| Store the unopened product at 2 to 8 degreesC. Do not use past expiration date. Protect from light. |
| Flow Cytometry | |
| 1054C | |
| Flow Cytometry 10 uL/10^6 cells | |
| FOXP3 | |
| E. coli-derived recombinant human FoxP3 Met1-Leu71 | |
| 25 Tests | |
| 50943 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction