missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kininogen/Kininostatin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
209.00€ - 520.00€
Specifications
| Antigen | Kininogen/Kininostatin/Bradykinin |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18717177
![]() |
R&D Systems
AF1396-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18053272
![]() |
R&D Systems
AF1396 |
100 μg |
520.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kininogen/Kininostatin/Bradykinin Polyclonal specifically detects Kininogen/Kininostatin/Bradykinin in Human samples. It is validated for Western Blot.Specifications
| Kininogen/Kininostatin/Bradykinin | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| alpha 2-thiol Proteinase Inhibitor, Alpha-2-thiol proteinase inhibitor, BDK, bradykinin, Fitzgerald factor, High molecular weight kininogen, kininogen, kininogen 1, kininogen-1, KNG1, KNGHMWK, Williams-Fitzgerald-Flaujeac factor | |
| KNG1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P01042 | |
| 3827 | |
| E. coli-derived recombinant human Kininostatin Lys438-Ser531 Accession # P01042 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Kininogen/Kininostatin in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title