Learn More
Human Kininogen/Kininostatin Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF1396-SP
This item is not returnable.
View return policy
Description
Kininogen/Kininostatin/Bradykinin Polyclonal specifically detects Kininogen/Kininostatin/Bradykinin in Human samples. It is validated for Western Blot.
Specifications
| Kininogen/Kininostatin/Bradykinin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha 2-thiol Proteinase Inhibitor, Alpha-2-thiol proteinase inhibitor, BDK, bradykinin, Fitzgerald factor, High molecular weight kininogen, kininogen, kininogen 1, kininogen-1, KNG1, KNGHMWK, Williams-Fitzgerald-Flaujeac factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3827 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P01042 | |
| KNG1 | |
| E. coli-derived recombinant human Kininostatin Lys438-Ser531 Accession # P01042 | |
| 25 μg | |
| Primary | |
| Detects human Kininogen/Kininostatin in direct ELISAs and Western blots. | |
| Human | |
| IgG |