missing translation for 'onlineSavingsMsg'
Learn More

Human IgG4 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

185.00€ - 482.00€

Specifications

Antigen Human IgG4
Clone 2350A
Dilution Western Blot 2 μg;/mL, ELISA
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18711977
GREENER_CHOICE
View Documents
R&D Systems
MAB98951-SP
25 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
18702217
GREENER_CHOICE
View Documents
R&D Systems
MAB98951-100
100 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Human IgG4 Monoclonal specifically detects Human IgG4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Human IgG4
Western Blot 2 μg;/mL, ELISA
Monoclonal
Supernatant
Human
Ig gamma-4 chain C region, immunoglobulin heavy constant gamma 4 (G4m marker), MGC117419
Synthetic peptide containing Human IgG4
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human IgG4 in Western blots and ELISA. In Western blots, no cross-reactivity with human IgG1, human IgG2, and human IgG3.
2350A
Western Blot, ELISA
Unconjugated
Rabbit
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
3503.0
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.