missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB98951-100
This item is not returnable.
View return policy
Description
Human IgG4 Monoclonal specifically detects Human IgG4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Human IgG4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human IgG4 in Western blots and ELISA. In Western blots, no cross-reactivity with human IgG1, human IgG2, and human IgG3. | |
| Human | |
| Supernatant |
| Western Blot, ELISA | |
| 2350A | |
| Western Blot 2 μg;/mL, ELISA | |
| Ig gamma-4 chain C region, immunoglobulin heavy constant gamma 4 (G4m marker), MGC117419 | |
| Synthetic peptide containing Human IgG4 | |
| 100 μg | |
| 3503.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction