missing translation for 'onlineSavingsMsg'
Learn More

Human GFR alpha-like Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

206.00€ - 460.00€

Specifications

Antigen GFR alpha-like
Clone 2345C
Dilution Flow Cytometry 0.25 μg/106 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready
Applications Flow Cytometry, Blocking Assay, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18710157
GREENER_CHOICE
View Documents
R&D Systems
MAB9697-SP
25 μg
206.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18717989
GREENER_CHOICE
View Documents
R&D Systems
MAB9697-100
100 μg
460.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GFR alpha-like Monoclonal specifically detects GFR alpha-like in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Specifications

Specifications

GFR alpha-like
Flow Cytometry 0.25 μg/106 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready
Monoclonal
Supernatant
Human
Q6UXV0
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
2345C
Flow Cytometry, Blocking Assay, CyTOF
Unconjugated
Rabbit
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Human embryonic kidney cell line HEK293-derived recombinant human GFR alpha-like, Ser19-Glu351, Accession # Q6UXV0
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human GFR alpha-like in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.