Learn More
Human GFR alpha-like Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB9697-SP
 View more versions of this product
Product Code. 18710157
  • 206.00€ / 25µg
Description
GFR alpha-like Monoclonal specifically detects GFR alpha-like in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GFR alpha-like
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Rabbit
Protein A or G purified from cell culture supernatant
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG
Flow Cytometry, Blocking Assay, CyTOF
2345C
Flow Cytometry 0.25 μg/106 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready
Q6UXV0
Human embryonic kidney cell line HEK293-derived recombinant human GFR alpha-like, Ser19-Glu351, Accession # Q6UXV0
25 μg
Detects human GFR alpha-like in direct ELISAs.
Human
Supernatant