missing translation for 'onlineSavingsMsg'
Learn More

Human Dopamine D2 R/DRD2 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

210.00€ - 425.00€

Specifications

Antigen Dopamine D2R/DRD2
Clone 960710
Dilution Immunohistochemistry 5-25 ug/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18758668
GREENER_CHOICE
View Documents
R&D Systems
MAB9266-SP
25 μg
210.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18742398
GREENER_CHOICE
View Documents
R&D Systems
MAB9266-100
100 μg
425.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dopamine D2R/DRD2 Monoclonal specifically detects Dopamine D2R/DRD2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Dopamine D2R/DRD2
Immunohistochemistry 5-25 ug/mL
Monoclonal
Mouse
Human
P14416
1813
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
960710
Immunohistochemistry
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, Dopamine D2R, dopamine receptor D2, dopamine receptor D2 isoform, DRD2, seven transmembrane helix receptor
Human DRD2 synthetic peptide Accession # P14416
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Dopamine D2 R/DRD2 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.