missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dopamine D2 R/DRD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9266-100
This item is not returnable.
View return policy
Description
Dopamine D2R/DRD2 Monoclonal specifically detects Dopamine D2R/DRD2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Dopamine D2R/DRD2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, Dopamine D2R, dopamine receptor D2, dopamine receptor D2 isoform, DRD2, seven transmembrane helix receptor | |
| Human DRD2 synthetic peptide Accession # P14416 | |
| 100 μg | |
| Primary | |
| Detects human Dopamine D2 R/DRD2 in direct ELISAs. | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| 960710 | |
| Immunohistochemistry 5-25 ug/mL | |
| P14416 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1813 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction