missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL26/Eotaxin-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
211.00€ - 1830.00€
Specifications
| Antigen | CCL26/Eotaxin-3 |
|---|---|
| Clone | 115002 |
| Dilution | Western Blot 2 ug/mL, Neutralization 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783878
![]() |
R&D Systems
MAB653-SP |
25 μg |
211.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18795779
![]() |
R&D Systems
MAB653-100 |
100 μg |
461.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18721387
![]() |
R&D Systems
MAB653-500 |
500 μg |
1830.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL26/Eotaxin-3 Monoclonal specifically detects CCL26/Eotaxin-3 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CCL26/Eotaxin-3 | |
| Western Blot 2 ug/mL, Neutralization 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q9Y258 | |
| 10344 | |
| E. coli-derived recombinant human CCL26/Eotaxin-3 Thr24-Leu94 Accession # Q9Y258 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CCL26/Eotaxin-3 in ELISAs and Western blots. Does not cross-react with recombinant human (rh) CCL11/Eotaxin, recombinant mouse (rm) CCL11/Eotaxin, rhCCL24/Eotaxin-2, rmCCL24/Eotaxin-2, rhCCL3, rhCCL4, rmCCL3, rmCCL4, rhCCL5, rhCCL13, or rhCCL14. |
| 115002 | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CC chemokine IMAC, C-C motif chemokine 26, chemokine (C-C motif) ligand 26, chemokine N1, eotaxin-3, Eotaxin3, IMAC, Macrophage inflammatory protein 4-alpha, MGC126714, MIP-4a, MIP-4alpha, SCYA26, SCYA26MIP-4-alpha, small inducible cytokine A26, small inducible cytokine subfamily A (Cys-Cys), member 26, Small-inducible cytokine A26, TSC-1Thymic stroma chemokine-1 | |
| CCL26 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title