missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL26/Eotaxin-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB653-500
This item is not returnable.
View return policy
Description
CCL26/Eotaxin-3 Monoclonal specifically detects CCL26/Eotaxin-3 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL26/Eotaxin-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CC chemokine IMAC, C-C motif chemokine 26, chemokine (C-C motif) ligand 26, chemokine N1, eotaxin-3, Eotaxin3, IMAC, Macrophage inflammatory protein 4-alpha, MGC126714, MIP-4a, MIP-4alpha, SCYA26, SCYA26MIP-4-alpha, small inducible cytokine A26, small inducible cytokine subfamily A (Cys-Cys), member 26, Small-inducible cytokine A26, TSC-1Thymic stroma chemokine-1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 10344 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| 115002 | |
| Western Blot 2 ug/mL, Neutralization 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9Y258 | |
| CCL26 | |
| E. coli-derived recombinant human CCL26/Eotaxin-3 Thr24-Leu94 Accession # Q9Y258 | |
| 500 μg | |
| Primary | |
| Detects human CCL26/Eotaxin-3 in ELISAs and Western blots. Does not cross-react with recombinant human (rh) CCL11/Eotaxin, recombinant mouse (rm) CCL11/Eotaxin, rhCCL24/Eotaxin-2, rmCCL24/Eotaxin-2, rhCCL3, rhCCL4, rmCCL3, rmCCL4, rhCCL5, rhCCL13, or rhCCL14. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction