missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1, p Ser326 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
155.00€ - 832.00€
Specifications
| Antigen | HSF1 (p Ser326) |
|---|---|
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18075389
|
Novus Biologicals
NBP1-97452-0.025MG |
0.025 mg |
155.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18007189
|
Novus Biologicals
NBP1-97452-0.05MG |
0.05 mg |
375.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18056339
|
Novus Biologicals
NBP1-97452-0.2MG |
0.2 mg |
832.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSF1 Polyclonal specifically detects HSF1 in Human samples. It is validated for Western Blot.Specifications
| HSF1 (p Ser326) | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| 3297 | |
| Synthetic peptide corresponding to the N-terminus of human HSF1 phosphorylated at Ser 326. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| HSF1 | |
| IgG | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title