missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1, p Ser326 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97452-0.025MG
This item is not returnable.
View return policy
Description
HSF1 Polyclonal specifically detects HSF1 in Human samples. It is validated for Western Blot.
Specifications
| HSF1 (p Ser326) | |
| Polyclonal | |
| Western Blot 1:1000 | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| HSF1 | |
| Synthetic peptide corresponding to the N-terminus of human HSF1 phosphorylated at Ser 326. | |
| 0.025 mg | |
| Phospho Specific | |
| 3297 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction