missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1 Antibody (10H8), Novus Biologicals™
Rat Monoclonal Antibody
175.00€ - 647.00€
Especificaciones
| Antigen | HSF1 |
|---|---|
| Clone | 10H8 |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|
18080149
|
Novus Biologicals
NBP1-97504-0.025MG |
0.025 mg |
175.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18018377
|
Novus Biologicals
NBP1-97504-0.05MG |
0.05 mg |
391.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18024169
|
Novus Biologicals
NBP1-97504-0.2MG |
0.2 mg |
647.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Especificaciones
| HSF1 | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| 3297 | |
| Recombinant mouse HSF1. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 10H8 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rat | |
| Rabbit, Rat, Primate, Human, Mouse | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| HSF1 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto