missing translation for 'onlineSavingsMsg'
Learn More

HSF1 Antibody (10H8), Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NBP1-97504-0.2MG

 View more versions of this product

Product Code. 18024169

  • 647.00€ / 0.20mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSF1
Monoclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
HSF1
Recombinant mouse HSF1.
0.2 mg
Primary
Human, Mouse, Rat, Primate, Rabbit
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
10H8
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Rat
Protein G purified
RUO
3297
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only