missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD9 Antibody (OTI1H3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45002-100ul
This item is not returnable.
View return policy
Description
HOXD9 Monoclonal antibody specifically detects HOXD9 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| HOXD9 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 3235 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI1H3 | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| homeo box D9, homeobox D9, Homeobox protein Hox-4C, Homeobox protein Hox-5.2, homeobox protein Hox-D9, HOX4, Hox-4.3, mouse, homolog of, HOX4CHox-4.3, Hox-5.2 | |
| Human recombinant protein fragment corresponding to amino acids 109-352 of human HOXD9 (NP_055028) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction