missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
224.00€ - 530.00€
Specifications
| Antigen | HOXB1 |
|---|---|
| Dilution | Western Blot 1:2000 - 1:4000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228272
|
Novus Biologicals
NBP3-33485-100ul |
100 μL |
530.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229312
|
Novus Biologicals
NBP3-33485-20ul |
20 μL |
224.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXB1 Monoclonal antibody specifically detects HOXB1 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| HOXB1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| homeo box 2I, homeo box B1, homeobox B1, Homeobox protein Hox-2I, homeobox protein Hox-B1, HOX2, Hox-2.9, HOX2IMGC116844, MGC116843, MGC116845 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 35-180 of human HOXB1 (NP_002135.2).,, Sequence:, SAQAVDSYASEGRYGGGLSSPAFQQNSGYPAQQPPSTLGVPFPSSAPSGYAPAACSPSYGPSQYYPLGQSEGDGGYFHPSSYGAQLGGLSDGYGAGGAGPGPYPPQHPPYGNEQTASFAPAYADLLSEDKETPCPSEPNTPTARTF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:2000 - 1:4000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 3211 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title