missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33485-100ul
This item is not returnable.
View return policy
Description
HOXB1 Monoclonal antibody specifically detects HOXB1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| HOXB1 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:4000, ELISA Recommended starting concentration is 1 μg/mL | |
| homeo box 2I, homeo box B1, homeobox B1, Homeobox protein Hox-2I, homeobox protein Hox-B1, HOX2, Hox-2.9, HOX2IMGC116844, MGC116843, MGC116845 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 35-180 of human HOXB1 (NP_002135.2).,, Sequence:, SAQAVDSYASEGRYGGGLSSPAFQQNSGYPAQQPPSTLGVPFPSSAPSGYAPAACSPSYGPSQYYPLGQSEGDGGYFHPSSYGAQLGGLSDGYGAGGAGPGPYPPQHPPYGNEQTASFAPAYADLLSEDKETPCPSEPNTPTARTF | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3211 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction