Learn More
HOXA10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
231.00€ - 326.00€
Specifications
| Antigen | HOXA10 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:2500-1:10000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
18610968
|
Novus Biologicals
NBP3-06103-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696887
|
Novus Biologicals
NBP3-06103-50ul |
50 μg |
231.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXA10 Polyclonal antibody specifically detects HOXA10 in Human samples. It is validated for Immunohistochemistry (Paraffin)Spécification
| HOXA10 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| homeo box A10, homeobox A10, homeobox protein 1H, Homeobox protein Hox-1.8, Homeobox protein Hox-1H, homeobox protein Hox-A10, homeobox protein HOXA10, HOX1, HOX1HHOX1.8, MGC12859, PL | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HOXA10. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:2500-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 3206 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit