missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06103-100ul
This item is not returnable.
View return policy
Description
HOXA10 Polyclonal antibody specifically detects HOXA10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HOXA10 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:2500-1:10000 | |
| homeo box A10, homeobox A10, homeobox protein 1H, Homeobox protein Hox-1.8, Homeobox protein Hox-1H, homeobox protein Hox-A10, homeobox protein HOXA10, HOX1, HOX1HHOX1.8, MGC12859, PL | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HOXA10. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 3206 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction