missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DRB3 Mouse anti-Human, Canine, Chimpanzee, Monkey, Baboon, Squirrel, Clone: Bra30, Novus Biologicals™
Mouse Monoclonal Antibody
519.00€
Specifications
| Antigen | HLA DRB3 |
|---|---|
| Clone | Bra30 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Flow Cytometry, Immunofluorescence |
Beskrivning
HLA DRB3 Monoclonal specifically detects HLA DRB3 in Human, Canine, Chimpanzee, Monkey, Baboon, Squirrel samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifikationer
| HLA DRB3 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Asthma, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| 3125 | |
| IgG2a κ | |
| Protein A or G purified |
| Bra30 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Canine, Chimpanzee, Monkey, Baboon | |
| DR7, HLA class II histocompatibility antigen, DR beta 3 chain, HLA class II histocompatibility antigen, DRB1-7 beta chain, HLA-DR3B, HLA-DR52, HLA-DRB1, human leucocyte antigen DRB3, major histocompatibility complex, class II, DR beta 3, MGC117330, MHC class II antigen DR beta 3 chain, MHC class II antigen DRB3, MHC class II HLA-DR beta 3 chain | |
| Human REH cells (non-T, non-B leukemia cell line) | |
| Primary | |
| Store at 4°C. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel