missing translation for 'onlineSavingsMsg'
Learn More

HLA DRB3 Mouse anti-Human, Canine, Chimpanzee, Monkey, Baboon, Squirrel, Clone: Bra30, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07168-100ug

 View more versions of this product

Product Code. 18694719

  • 519.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HLA DRB3 Monoclonal specifically detects HLA DRB3 in Human, Canine, Chimpanzee, Monkey, Baboon, Squirrel samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA DRB3
Monoclonal
0.2 mg/mL
Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL
DR7, HLA class II histocompatibility antigen, DR beta 3 chain, HLA class II histocompatibility antigen, DRB1-7 beta chain, HLA-DR3B, HLA-DR52, HLA-DRB1, human leucocyte antigen DRB3, major histocompatibility complex, class II, DR beta 3, MGC117330, MHC class II antigen DR beta 3 chain, MHC class II antigen DRB3, MHC class II HLA-DR beta 3 chain
Human REH cells (non-T, non-B leukemia cell line)
100 μg
Asthma, Immunology
3125
Store at 4°C.
IgG2a κ
Flow Cytometry, Immunofluorescence
Bra30
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human, Canine, Chimpanzee, Monkey, Baboon
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.